Buffered oxide etch (7:1) VLSI for microelectronic use, J.T.Baker®

Danger

5361-03EA 0 KRW
5361-03
Buffered oxide etch (7:1) VLSI for microelectronic use, J.T.Baker®
Buffered oxide etch
UN: 2922
ADR: 8,II

Order Now

사양 테스트 결과

For Microelectronic Use
Ship and store material above 55°F (13°C)
Ammonium Fluoride (NH₄F)(w/w) 34.3 - 35.3 %
Hydrofluoric Acid (HF)(w/w) 6.18 - 6.48 %
Residue after Ignition ≤10 ppm
Trace Impurities - Aluminum (Al) ≤200.0 ppb
Arsenic and Antimony (as As) ≤30.0 ppb
Trace Impurities - Barium (Ba) ≤1000.0 ppb
Trace Impurities - Boron (B) ≤200.0 ppb
Trace Impurities - Cadmium (Cd) ≤500.0 ppb
Trace Impurities - Calcium (Ca) ≤200.0 ppb
Trace Impurities - Chromium (Cr) ≤100.0 ppb
Trace Impurities - Cobalt (Co) ≤500.0 ppb
Trace Impurities - Copper (Cu) ≤100.0 ppb
Trace Impurities - Gallium (Ga) ≤500.0 ppb
Trace Impurities - Germanium (Ge) ≤500.0 ppb
Trace Impurities - Gold (Au) ≤300.0 ppb
Heavy Metals (as Pb) ≤1000.0 ppb
Trace Impurities - Iron (Fe) ≤200.0 ppb
Trace Impurities - Lead (Pb) ≤300.0 ppb
Trace Impurities - Lithium (Li) ≤1000.0 ppb
Trace Impurities - Magnesium (Mg) ≤200.0 ppb
Trace Impurities - Manganese (Mn) ≤200.0 ppb
Trace Impurities - Nickel (Ni) ≤200.0 ppb
Trace Impurities - Potassium (K) ≤300.0 ppb
Trace Impurities - Silver (Ag) ≤500.0 ppb
Trace Impurities - Sodium (Na) ≤300 ppb
Trace Impurities - Strontium (Sr) ≤1000.0 ppb
Trace Impurities - Tin (Sn) ≤300.0 ppb
Trace Impurities - Titanium (Ti) ≤300.0 ppb
Trace Impurities - Zinc (Zn) ≤300.0 ppb
Particle Count - 1.0 µm and greater ≤25 par/ml
Reported value is the average of all samples counted for this lot number
with no individual sample value exceeding the specification.
For additional information, go to www.askavantor.com. Search keywords
freezing" and product name."

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR