Tetrabutylammonium hydroxide 1 M in methanol, BAKER ANALYZED® for ion pair chromatography, for HPLC, J.T.Baker®


Filtered through a 0.2 µm filter.

V362-05EA 0 KRW
V362-05
Tetrabutylammonium hydroxide 1 M in methanol, BAKER ANALYZED® for ion pair chromatography, for HPLC, J.T.Baker®
Tetrabutylammonium hydroxide
포뮬러: C₁₆H₃₇NO
MW: 259,48 g/mol
녹는점: 29.85 °C
보관 온도: 2 - 8°C
MDL 번호: MFCD00009425
CAS 번호: 2052-49-5
EINECS: 218-147-6
UN: 2924
ADR: 3,II

Order Now

사양 테스트 결과

For Laboratory,Research,or Manufacturing Use
Molarity 0.98 - 1.08
1.0 M Tris Base
1.0 M Acetate
0.025 M EDTA
Filtered through a 0.2 micron filter.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR