D-(+)-Biotin, J.T.Baker®

동의어: Vitamin HVitamin B8D-BiotinVitamin B7

C272-00EA 0 KRW
C272-00 C272-05
D-(+)-Biotin, J.T.Baker®
D-(+)-Biotin
포뮬러: C₁₀H₁₆N₂O₃S
MW: 244,31 g/mol
녹는점: 232…233 °C
밀도: 1,41 g/cm³ (20 °C)
보관 온도: Refrigerator
MDL 번호: MFCD00005541
CAS 번호: 58-85-5
EINECS: 200-399-3
UN: 0000
Merck 색인: 13,01231

Order Now

사양 테스트 결과

For Laboratory,Research,or Manufacturing Use
Melting Point 229 - 232 °C
Specific Rotation, [Å]²⁵^D (c=1 in 0.1N NaOH) (+) 89 - 93 Degree

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR