Ethanol denatured ≥99.5%, GPR RECTAPUR® (denatured with 1% MEK)
Danger
85033.550EA 0 KRW
85033.550 85033.320 85033.460 85033.360
Ethanol denatured ≥99.5%, GPR RECTAPUR® (denatured with 1% MEK)
Ethanol denatured
포뮬러: H₃CCH₂OH
MW: 46,07 g/mol
밀도: 0,79 g/cm³ (20 °C)
보관 온도: Ambient
MDL 번호: MFCD00003568
CAS 번호: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3,II
Order Now
사양 테스트 결과
Assay (GC) Min. 99.3 %
Assay (Ethanol) (V/V) Min. 98.3 %
Appearance Clear colourless liquid
Identification (IR) Passes test
Density (20/4) 0.780 - 0.800
Ethyl isoamyl ketone Min. 150 ppm
Methyl ethyl ketone (V/V) 0.8 - 1.3 %
Methyl isopropyl ketone Min. 250 ppm
Evaporation residue Max. 50 ppm
Water Max. 0.3 %
Denat. BrStV §44 ,1a Conform