PE gloves
공급업체: VWR BRAND (P/L-PRODUCTS USA)
VWR®
VWRU32915-224EA 1341.8 KRW
VWRU32915-224 VWRU32915-246 VWRU32915-268
PE gloves
Gloves
These single-use polyethylene gloves with extra-long cuffs provide excellent resistance to chemicals and detergents.
  • Ambidextrous
  • Embossed finish for improved grip
  • Tough, waterproof construction
  • Soft, virgin polyethylene gloves are so pliable that you can handle even small laboratory apparatus with ease
  • Convenient pull-out dispenser
Ideal for hand and product protection in virtually any type of laboratory, food handling, industrial safety, veterinary, electronics, painting, printing, or photography environments.