Cleanroom gloves, nitrile, CERTICLEAN®
공급업체: VWR BRAND (P/L-PRODUCTS USA)
CERTICLEAN®
VWRU40101-220EA 3933.9 KRW InStock
VWRU40101-220 VWRU40101-226 VWRU40101-228 VWRU40101-224 VWRU40101-222
Cleanroom gloves, nitrile, CERTICLEAN®
Gloves
Ambidextrous, powder-free gloves with beaded cuff for strength and reduction of cuff roll down. Strong, puncture-resistant construction.
  • Non sterile, powder-free
  • Film thickness designed for comfort and tactile sensitivity
  • Textured fingertips for a reliable grip
  • Manufacturing processes deliver low particle counts
  • Reduced ionic contaminants and nonvolatile residue (NVR) transfer
  • In-use statistic dissipative properties superior to natural rubber latex
Suitable for use in Class 100, ISO 5, M3.5 and lower cleanrooms.
포장: Packaged in cleanroom-compatible polyethylene, 100 gloves/double poly bag.
Order Now
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Learn more