Antibacterial hand cleansers, SoftCIDE™
공급업체: VWR Collection
VWR® SoftCIDE®
VWRU56614-420EA 225270 KRW
VWRU56614-420 VWRU56700-124 VWRU56614-424
Antibacterial hand cleansers, SoftCIDE™
Skin Care Soaps and Skin Cleansers
Extra-mild, fast-acting, antimicrobial soap with 1% PCMX. Specialised formulations and mild ingredients help to replenish the natural oils of the skin.
  • Effective against a broad spectrum micro-organism
  • pH balanced with the skin
  • Triclosan-free