N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride, high purity

공급업체: VWR Chemicals
Warning

N195-5GEA 0 KRW InStock
N195-5G N195-25G
N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride, high purity
N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride
포뮬러: C₈H₁₇N₃·HCl
MW: 191,7 g/mol
녹는점: 112-113 °C
보관 온도: Freezer
MDL 번호: MFCD00012503
CAS 번호: 25952-53-8
EINECS: 247-361-2
UN: 0000
https://kr.vwr.com/stibo/web/std.lang.all/14/32/20961432.jpg

Order Now

사양 테스트 결과

Carbon 49.7 - 50.7 %
Appearance White powder
Nitrogen 21.4 - 22.4 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more