1,3-Dimethylurea ≥96.5%

공급업체: VWR Chemicals

9705-500GEA 990.7 KRW InStock
9705-500G 9705-1KG
1,3-Dimethylurea ≥96.5%
1,3-Dimethylurea
포뮬러: C₃H₈N₂O
MW: 88,11 g/mol
끊는점: 268 °C (1013 hPa)
녹는점: 96 °C
밀도: 1,142 g/cm³ (20 °C)
보관 온도: Ambient
MDL 번호: MFCD00008286
CAS 번호: 96-31-1
EINECS: 202-498-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more