TMB Liquid membrane substrate for biotechnology, VWR
공급업체: Avantor
VWRCE888-100MLEA 863900 KRW
VWRCE888-100ML
TMB Liquid membrane substrate for biotechnology, VWR
TMB (3,3',5,5'-tetramethylbenzidine) Membrane Substrate consists of two solutions that are mixed immediately before use for the detection of horseradish peroxidase (HRP)-labeled probes in membrane- and immunohistochemical-based assays.
This chromogenic substrate, in the presence of HRP, yields an insoluble blue product that may be observed with ambient light. The TMB Membrane Substrate solutions, when mixed, are sufficient for detection of 500 cm² of membrane.
Store cold (4 - 8 °C).
주의: For research use only. Not for therapeutic or diagnostic use.