Alcian blue

Warning

동의어: C.I. 74240C.N. AnAn-6Ingrain Blue 1Alcian blue 8 GX

0298-50GEA 3725730 KRW
0298-100G 0298-50G
Alcian blue
Alcian blue

Em (610 nm, water)

포뮬러: C₅₆H₆₈Cl₄CuN₁₆S₄
MW: 1298,86 g/mol
보관 온도: Ambient
MDL 번호: MFCD00010720
CAS 번호: 33864-99-2
EINECS: 251-705-7

Order Now

사양 테스트 결과

Em (610 nm, water) >26000

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR