VWR®, Extraction solvent stock solution in bottles
공급업체: Avantor
VWRC87508.004EA 481110 KRW
VWRC87508.004
VWR®, Extraction solvent stock solution in bottles
Mixture of C10 (20 μL/L) and C40 (20 mg/L). Solvent : Hexane - petroleum ether (1:1).
Packed in 1,5 and 4,5 ml Certan® bottles
001 = 1×1,5 ml
002 = 1×4,5 ml
003 = 3×1,5 ml
004 = 3×4,5 ml