4-Nitrophenyl phosphate disodium salt hexahydrate (pNPP) substrate 30 mg, tablets, Reagent Grade

공급업체: VWR Chemicals
Warning

0617-1KTEA 12922.5 KRW InStock
0617-1KT 0617-100TABS
4-Nitrophenyl phosphate disodium salt hexahydrate (pNPP) substrate 30 mg, tablets, Reagent Grade
4-Nitrophenyl phosphate disodium salt hexahydrate (pNPP)

Preferred substrate for high sensitivity detection of alkaline phosphatase in EIA assays. Read at 405 nm. Each 175 mg tablet contains 30 mg of p-NPP.


Reaction condition: 1 mg/ml in 10 mM diethanolamine and 0.5 mM MgCl2, pH 9,5

포뮬러: O₂NC₆H₄OP(O)(ONa)₂·6H₂O
MW: 371,15 g/mol
녹는점: 300 °C
보관 온도: -20°C
MDL 번호: MFCD00066288
CAS 번호: 4264-83-9
EINECS: 224-246-5
UN: 0000

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more