Standard mixture of 4 n-alkanes in Certan® bottles according to ISO9377-2
공급업체: VWR Chemicals
87506.001EA 3233.9 KRW InStock
87506.001 87506.004 87506.002 87506.003
Standard mixture of 4 n-alkanes in Certan® bottles according to ISO9377-2
Standards Organic Standards
C10+C20+C30+C40
Concentration : 100 mg/l each
Solvent : Hexane - petroleum ether (1:1)