Surface cleaning and disinfectant products, Pursept® AF
공급업체: Brand
BRND44855EA 0 KRW
BRND44850_B BRND44855
Surface cleaning and disinfectant products, Pursept® AF
세정제 살균제
A disinfecting detergent for cleaning and disinfection of floors, walls and fittings in one operation.
  • Appropriate for use in the food sector
  • Clear, green liquid with fresh smell
  • Mildly alkaline of 1% solution with pH value of approximately 10
As recommended by the RKI, Bundesgesundheitsblatt 01/2004.