Sodium hydroxide 0.01 mol/l (0.01 N) in aqueous solution

공급업체: VWR Chemicals
Danger

305289.1000EA 379.1 KRW InStock
305289.1000
Sodium hydroxide 0.01 mol/l (0.01 N) in aqueous solution
Sodium hydroxide
포뮬러: NaOH
MW: 40 g/mol
끊는점: 1390 °C (1013 hPa)
녹는점: 323 °C
밀도: 1 g/cm³ (25 °C)
보관 온도: Ambient
MDL 번호: MFCD00003548
CAS 번호: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,III
REACH: 01-2119457892-27
Merck 색인: 14,08627

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more