Bromothymol blue, BAKER ANALYZED® ACS, J.T.Baker®
Warning
|
동의어:
3′,3″-Dibromothymolsulphonphthalein
D470-03EA
954600
KRW
D470-03
Bromothymol blue, BAKER ANALYZED® ACS, J.T.Baker®
Bromothymol blue
Lot analysis on label.
포뮬러:
C₂₇H₂₈Br₂O₅S MW: 624,39 g/mol 녹는점: 204…202 °C 보관 온도: Ambient |
MDL 번호:
MFCD00005872 CAS 번호: 76-59-5 EINECS: 200-971-2 UN: 0000 Merck 색인: 13,01430 |
사양 테스트 결과
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
ACS - Clarity of Solution | Passes Test |
Visual Transition Interval - pH…7.6 (Blue) | Passes Test |
Visual Transition Interval - pH…6.7 (Green) | Passes Test |
Visual Transition Interval - pH…6.0 (Yellow) | Passes Test |
Alkali Consumption | Passes Test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...