BAKERBOND spe™ columns are disposable PP or glass columns pre-packed with high capacity sorbents contained between two 20 µm PE frits.
- Silica based
- Extraction of compounds from sample solution volumes ranging from a few hundred µl up to 1 L
- Processing by vacuum, positive pressure or centrifugation
General applications: Small peptides, separations where C18 gives excessive retention or poor recovery.
These SPE columns can be used in reversed phase mode.
포장: Certain columns are available in a foil-lined fibre drum called the Jumbo Pack. The columns are loose packed in the foil liner to eliminate the inconvenience of opening individual pouches.