J.T.Baker® BAKERBOND spe™ WP Butyl (C4), SPE Columns
공급업체: Avantor
J.T.Baker® BAKERBOND spe™
7216-06EA 6114000 KRW
7216-06
J.T.Baker® BAKERBOND spe™ WP Butyl (C4), SPE Columns
크로마토그래피 컬럼
BAKERBOND spe™ columns are disposable PP or glass columns pre-packed with high capacity sorbents contained between two 20 µm PE frits.
  • Silica based
  • Extraction of compounds from sample solution volumes ranging from a few hundred µl up to 1 L
  • Processing by vacuum, positive pressure or centrifugation
General applications: Small peptides, separations where C18 gives excessive retention or poor recovery.
These SPE columns can be used in reversed phase mode.
포장: Certain columns are available in a foil-lined fibre drum called the Jumbo Pack. The columns are loose packed in the foil liner to eliminate the inconvenience of opening individual pouches.