Arsenite solution Reag. Ph. Eur. 1008301
공급업체: VWR Chemicals
87781.180EA 1736.3 KRW InStock
87781.180
Arsenite solution Reag. Ph. Eur. 1008301
Arsenite solution
Composition: 0.5% arsenite, 2.0% sodium hydrogen carbonate
주문 정보: 주문 정보: 주문 정보: 주문 정보: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.
EINECS: 000-000-0