Potassium fluoride, BAKER ANALYZED® ACS, J.T. Baker®
공급업체: AVANTOR PERFORMANCE MATERIALS
Danger
3123-05EA 0 KRW InStock
3123-05 3123-01
Potassium fluoride, BAKER ANALYZED® ACS, J.T. Baker®
Potassium fluoride
포뮬러: KF
MW: 58.1 g/mol
끊는점: 1505 °C (1013 hPa)
녹는점: 860 °C
밀도: 2,48 g/cm³ (20 °C)
보관 온도: Ambient
MDL 번호: MFCD00011398
CAS 번호: 7789-23-3
EINECS: 232-151-5
UN: 1812
ADR: 6.1,III
Merck 색인: 12,07794
Order Now
사양 테스트 결과
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (KF) ≥ 99.0 %
ACS - Chloride (Cl) ≤ 0.005 %
ACS - Titrable Acid (meq/g) ≤ 0.03
ACS - Titrable Base (meq/g) ≤ 0.01
ACS - Potassium Fluosilicate (K₂SiF₆) ≤ 0.1 %
ACS - Sulfate (SO₄) ≤ 0.005 %
ACS - Heavy Metals (as Pb) ≤ 0.001 %
ACS - Iron (Fe) ≤ 0.001 %
ACS - Sodium (Na) ≤ 0.2 %