L(-)-Tyrosine 98.5-101.5% (dried basis) USP, Multi-compendial, J.T. Baker®
공급업체: Avantor
동의어: TyrosineL-Tyrosine(S)-(-)-TyrosineH-Tyr-OH
2093-06EA 0 KRW
2093-06
L(-)-Tyrosine 98.5-101.5% (dried basis) USP, Multi-compendial, J.T. Baker®
L(-)-Tyrosine
  • Food GMP manufactured product
포뮬러: C₉H₁₁NO₃
MW: 181.19 g/mol
끊는점: 351 °C (1013 hPa)
녹는점: 285 °C
밀도: 1.41 g/cm³ (20 °C)
보관 온도: Ambient
MDL 번호: MFCD00002606
CAS 번호: 60-18-4
EINECS: 200-460-4
UN: 0000
Merck 색인: 12,09970
Order Now
사양 테스트 결과
Food GMP Manufactured Product
GMP Manufactured Product
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
USP - Assay (C₉H₁₁NO₃) (dried basis) 98.5 - 101.5 %
USP - Identification Passes Test
USP - Specific Rotation α²⁵D -11.2 - -9.8 Degree
USP - Loss on Drying ≤ 0.3 %
USP - Residue on Ignition ≤ 0.4 %
USP - Chloride (Cl) ≤ 0.04 %
USP - Sulfate (SO₄) ≤ 0.04 %
USP - Iron (Fe) ≤ 0.003 %
USP - Related Compounds - Individual Impurities ≤ 0.5 %
USP - Related Compounds - Total Impurities ≤ 2.0 %
EP/BP - Assay (C₉H₁₁NO₃) (dried basis) 99.0 - 101.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Specific Rotationα²ºD -12.3 - -11.0 Degree
EP/BP - Ninhydrin-Positive Substances - Impurity A ≤ 0.5 %
EP/BP-Ninhydrin-Positive Substances-Each ≤ 0.2 %
EP/BP - Ninhydrin Positive Substances - Total Impurities ≤ 0.6
EP/BP - Chloride (Cl) ≤ 200 ppm
EP/BP - Sulfate (SO₄) ≤ 300 ppm
EP/BP - Ammonium (NH₄) ≤ 0.02 %
EP/BP - Iron (Fe) ≤ 10 ppm
EP/BP - Loss on Drying ≤ 0.5 %
EP/BP - Ash (sulfated) ≤ 0.1 %
Endotoxin Concentration, IU/mg, For Information Only
Residual Solvents - No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
Elemental Impurities (USP <232>, EP 5.20) - information on elemental impurities for this product is available on the associated Product Regulatory Data Sheet and elemental impurity profile report.